accessibility menu, dialog, popup

UserName

CAS RN 2550-06-3

ClSiClClCl

CAS RN 2550-06-3

IUPAC Name: trichloro(3-chloropropyl)silane
Synonyms: trichloro(3-chloropropyl)silane
Molecular Weight (g/mol): 211.97
Molecular Formula: C3H6Cl4Si
InChi Key: OOXSLJBUMMHDKW-UHFFFAOYSA-N
SMILES: ClCCC[Si](Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results