CAS RN 24159-36-2
CAS RN 24159-36-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Norepinephrine Sulfonic Acid, TRC
CAS: 24159-36-2 Molecular Formula: C8 H11 N O5 S Molecular Weight (g/mol): 233.24 Synonym: α-(Aminomethyl)-3,4-dihydroxy-benzenemethanesulfonic Acid IUPAC Name: 2-amino-1-(3,4-dihydroxyphenyl)ethanesulfonic acid SMILES: NCC(c1ccc(O)c(O)c1)S(=O)(=O)O