accessibility menu, dialog, popup

UserName

CAS RN 2380-63-4

NH2NNHNN

CAS RN 2380-63-4

IUPAC Name: 1H-pyrazolo[3,4-d]pyrimidin-4-amine
Synonyms: 1H-pyrazolo[3,4-d]pyrimidin-4-amine
Molecular Weight (g/mol): 135.13
Molecular Formula: C5H5N5
InChi Key: LHCPRYRLDOSKHK-UHFFFAOYSA-N
SMILES: NC1=C2C=NNC2=NC=N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results