accessibility menu, dialog, popup

UserName

CAS RN 2373-31-1

CH3OOOH

CAS RN 2373-31-1

IUPAC Name: 6-hydroxy-4-methyl-2H-chromen-2-one
Synonyms: 6-hydroxy-4-methylchromen-2-one
Molecular Weight (g/mol): 176.17
Molecular Formula: C10H8O3
InChi Key: IRUHWRSITUYICV-UHFFFAOYSA-N
SMILES: CC1=CC(=O)OC2=CC=C(O)C=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results