accessibility menu, dialog, popup

UserName

OFFF

CAS RN 2367-02-4

IUPAC Name: 1-phenoxy-4-(trifluoromethyl)benzene
Synonyms: 1-phenoxy-4-(trifluoromethyl)benzene
Molecular Weight (g/mol): 238.21
Molecular Formula: C13H9F3O
InChi Key: UZJUDUZMPNCXPF-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=CC=C(OC2=CC=CC=C2)C=C1
Boiling Point 
  • (2)
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results