CAS RN 23646-68-6
CAS RN 23646-68-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
p-Nitrophenyl 2-Acetamido-2-deoxy-Alpha-D-galactopyranoside, TRC
CAS: 23646-68-6 Molecular Formula: C14 H18 N2 O8 Molecular Weight (g/mol): 342.3 IUPAC Name: N-[(2R,3R,4R,5R,6R)-4,5-dihydroxy-6-(hydroxymethyl)-2-(4-nitrophenoxy)oxan-3-yl]acetamide SMILES: CC(=O)N[C@@H]1[C@@H](O)[C@@H](O)[C@@H](CO)O[C@@H]1Oc2ccc(cc2)[N+](=O)[O-]