accessibility menu, dialog, popup

UserName

CAS RN 2362-64-3

H3CONH2S

CAS RN 2362-64-3

IUPAC Name: 4-methoxybenzene-1-carbothioamide
Synonyms: 4-methoxybenzenecarbothioamide
Molecular Weight (g/mol): 167.23
Molecular Formula: C8H9NOS
InChi Key: WKWVTPKUHJOVTI-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)C(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results