CAS RN 23299-18-5
IUPAC Name:
1-(3-chlorophenyl)-2-[(propan-2-yl)amino]ethan-1-ol
Synonyms:
1-(3-chlorophenyl)-2-(isopropylamino)ethanol
Molecular Weight (g/mol):
213.71
Molecular Formula:
C11H16ClNO
InChi Key:
WPRYXHDMUFXECU-UHFFFAOYNA-N
SMILES:
CC(C)NCC(O)C1=CC(Cl)=CC=C1
1
–
1
of
1
results
1
1-(3-Chlorophenyl)-2-(isopropylamino)ethanol, Thermo Scientific™
CAS: 23299-18-5 Molecular Formula: C11H16ClNO Molecular Weight (g/mol): 213.705 MDL Number: MFCD08060964 InChI Key: WPRYXHDMUFXECU-UHFFFAOYSA-N Synonym: 1-3-chlorophenyl-2-isopropylamino ethanol,1-3-chloro-phenyl-2-isopropylamino-ethanol,1-3-chlorophenyl-2-methylethyl amino ethan-1-ol,1-3-chlorophenyl-2-propan-2-yl amino ethan-1-ol,benzenemethanol,3-chloro-a-1-methylethyl amino methyl PubChem CID: 19913640 IUPAC Name: 1-(3-chlorophenyl)-2-(propan-2-ylamino)ethanol SMILES: CC(C)NCC(C1=CC(=CC=C1)Cl)O