accessibility menu, dialog, popup

UserName

CAS RN 230-27-3

N

CAS RN 230-27-3

IUPAC Name: benzo[h]quinoline
Synonyms: benzo(H)quinoline
Molecular Weight (g/mol): 179.22
Molecular Formula: C13H9N
InChi Key: WZJYKHNJTSNBHV-UHFFFAOYSA-N
SMILES: C1=CC=C2C(C=CC3=CC=CN=C23)=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)