accessibility menu, dialog, popup

UserName

CAS RN 22971-62-6

IUPAC Name: 5-oxo-5-(thiophen-2-yl)pentanoic acid
Synonyms: 5-oxo-5-(thiophen-2-yl)pentanoic acid
Molecular Weight (g/mol): 198.24
Molecular Formula: C9H10O3S
InChi Key: GSQMRVFOHNXKOR-UHFFFAOYSA-N
SMILES: OC(=O)CCCC(=O)C1=CC=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results