CAS RN 22934-51-6
CAS RN 22934-51-6
IUPAC Name:
2-hydroxy-3-methoxy-5-(prop-2-en-1-yl)benzaldehyde
Synonyms:
2-hydroxy-3-methoxy-5-(prop-2-en-1-yl)benzaldehyde
Molecular Weight (g/mol):
192.21
Molecular Formula:
C11H12O3
InChi Key:
FDHXEIOBIOVBEN-UHFFFAOYSA-N
SMILES:
COC1=CC(CC=C)=CC(C=O)=C1O
1
–
1
of
1
results
1
5-Allyl-2-hydroxy-3-methoxybenzaldehyde, 98%
CAS: 22934-51-6 Molecular Formula: C11H12O3 Molecular Weight (g/mol): 192.21 MDL Number: MFCD06408022 InChI Key: FDHXEIOBIOVBEN-UHFFFAOYSA-N Synonym: 5-allyl-2-hydroxy-3-methoxybenzaldehyde,5-allyl-3-methoxysalicylaldehyde,benzaldehyde, 2-hydroxy-3-methoxy-5-2-propenyl,benzaldehyde,2-hydroxy-3-methoxy-5-2-propen-1-yl,2-hydroxy-3-methoxy-5-prop-2-en-1-yl benzaldehyde,formyl eugenol,acmc-209fzq,2-hydroxy-3-methoxy-5-2-propenyl benzaldehyde,3-methoxy-2-oxidanyl-5-prop-2-enyl-benzaldehyde PubChem CID: 1713887 IUPAC Name: 2-hydroxy-3-methoxy-5-prop-2-enylbenzaldehyde SMILES: COC1=C(C(=CC(=C1)CC=C)C=O)O