accessibility menu, dialog, popup

UserName

(S)(R)OOHHOOHOHOH

CAS RN 225937-10-0

IUPAC Name: 2-(3,4-dihydroxyphenyl)-3,4-dihydro-2H-1-benzopyran-3,5,7-triol hydrate
Synonyms: (-)-catechol hydrate
Molecular Weight (g/mol): 308.29
Molecular Formula: C15H16O7
InChi Key: OFUMQWOJBVNKLR-UHFFFAOYNA-N
SMILES: O.OC1CC2=C(O)C=C(O)C=C2OC1C1=CC(O)=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)