CAS RN 2248003-60-1
CAS RN 2248003-60-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
AZD3229, MedChemExpress
MedChemExpress AZD3229 is a potent pan-KIT mutant inhibitor for the treatment of gastrointestinal stromal tumors.