accessibility menu, dialog, popup

UserName

H3COHCH3OHH3C

CAS RN 22422-34-0

IUPAC Name: 2,6,6-trimethylbicyclo[3.1.1]heptane-2,3-diol
Synonyms: pinanediol
Molecular Weight (g/mol): 170.25
Molecular Formula: C10H18O2
InChi Key: MOILFCKRQFQVFS-UHFFFAOYNA-N
SMILES: CC1(C)C2CC1C(C)(O)C(O)C2
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)
Boiling Point 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)