CAS RN 2240998-88-1
CAS RN 2240998-88-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
IEICO-4Cl, Thermo Scientific Chemicals
CAS: 2240998-88-1 Molecular Formula: C{1}{1}{4}H{1}{1}{4}Cl{4}N{4}O{4}S{4} Synonym: 2,2 '-((2 Z,2 'Z)-(((4,4,9-tris(4-hexylphenyl)-9-(4-pentylphenyl)-4,9-dihydro-s-indaceno[1,2-b:5,6-b dithiophene-2,7-diyl)bis(4-((2-ethylhexyl)oxy)thiophene5,2-diyl))bis(methanylylidene))bis(5,6-dichloro-3-oxo-2,3-dihydro-1 H-indene-2,1-diylidene))dimalononitrile