accessibility menu, dialog, popup

UserName

CAS RN 22282-99-1

CH3BrN

CAS RN 22282-99-1

IUPAC Name: 4-bromo-2-methylpyridine
Synonyms: 4-bromo-2-methylpyridine
Molecular Weight (g/mol): 172.03
Molecular Formula: C6H6BrN
InChi Key: JFBMFWHEXBLFCR-UHFFFAOYSA-N
SMILES: CC1=CC(Br)=CC=N1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results