accessibility menu, dialog, popup

UserName

CAS RN 22190-33-6

HNBr

CAS RN 22190-33-6

IUPAC Name: 5-bromo-2,3-dihydro-1H-indole
Synonyms: 5-bromo-2,3-dihydro-1H-indole
Molecular Weight (g/mol): 198.06
Molecular Formula: C8H8BrN
InChi Key: QEDCHCLHHGGYBT-UHFFFAOYSA-N
SMILES: BrC1=CC2=C(NCC2)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)