accessibility menu, dialog, popup

UserName

CAS RN 22174-29-4

IUPAC Name: 3-(2,4-dimethoxyphenyl)propanoic acid
Synonyms: 3-(2,4-dimethoxyphenyl)propanoic acid
Molecular Weight (g/mol): 210.23
Molecular Formula: C11H14O4
InChi Key: NVDCWSFMCUWKAL-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=C(CCC(O)=O)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results