CAS RN 221313-10-6
IUPAC Name:
methyl 6-(trifluoromethyl)pyridine-3-carboxylate
Synonyms:
methyl 6-(trifluoromethyl)pyridine-3-carboxylate
Molecular Weight (g/mol):
205.14
Molecular Formula:
C8H6F3NO2
InChi Key:
NEATXKNYTRXHQL-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CN=C(C=C1)C(F)(F)F
1
–
2
of
2
results
1
Methyl 6-(trifluoromethyl)nicotinate, 97%, Thermo Scientific™
CAS: 221313-10-6 Molecular Formula: C8H6F3NO2 Molecular Weight (g/mol): 205.136 MDL Number: MFCD03001346 InChI Key: NEATXKNYTRXHQL-UHFFFAOYSA-N PubChem CID: 2775647 IUPAC Name: methyl 6-(trifluoromethyl)pyridine-3-carboxylate SMILES: COC(=O)C1=CN=C(C=C1)C(F)(F)F
2
Methyl 6-(trifluoromethyl)nicotinate, 97%, Thermo Scientific™
CAS: 221313-10-6 Molecular Formula: C8H6F3NO2 Molecular Weight (g/mol): 205.13 MDL Number: MFCD03001346 InChI Key: NEATXKNYTRXHQL-UHFFFAOYSA-N PubChem CID: 2775647 IUPAC Name: methyl 6-(trifluoromethyl)pyridine-3-carboxylate SMILES: COC(=O)C1=CN=C(C=C1)C(F)(F)F