accessibility menu, dialog, popup

UserName

CAS RN 220513-46-2

NFBr

CAS RN 220513-46-2

IUPAC Name: 6-bromo-8-fluoroquinoline
Synonyms: 6-bromo-8-fluoroquinoline
Molecular Weight (g/mol): 226.05
Molecular Formula: C9H5BrFN
InChi Key: DDCOYWOYIUEOHQ-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=CC2=CC=CN=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results