accessibility menu, dialog, popup

UserName

CH3OOOHOH(E/Z)

CAS RN 220430-82-0

IUPAC Name: 3-(4-hydroxy-3-methoxyphenyl)-1-(2-hydroxyphenyl)prop-2-en-1-one
Synonyms: 3-(4-hydroxy-3-methoxyphenyl)-1-(2-hydroxyphenyl)prop-2-en-1-one
Molecular Weight (g/mol): 270.28
Molecular Formula: C16H14O4
InChi Key: NGQQEEYHKPKSER-UHFFFAOYSA-N
SMILES: COC1=C(O)C=CC(C=CC(=O)C2=CC=CC=C2O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results