CAS RN 2199-44-2
CAS RN 2199-44-2
IUPAC Name:
ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate
Synonyms:
ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate
Molecular Weight (g/mol):
167.21
Molecular Formula:
C9H13NO2
InChi Key:
IZSBSZYFPYIJDI-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(C)C=C(C)N1
1
–
1
of
1
results
1
Ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate, 98%, Thermo Scientific™
CAS: 2199-44-2 Molecular Formula: C9H13NO2 Molecular Weight (g/mol): 167.21 MDL Number: MFCD00010645 InChI Key: IZSBSZYFPYIJDI-UHFFFAOYSA-N Synonym: 3,5-dimethyl-1h-pyrrole-2-carboxylic acid ethyl ester,ethyl 3,5-dimethylpyrrole-2-carboxylate,ethyl 3,5-dimethyl-2-pyrrolecarboxylate,1h-pyrrole-2-carboxylic acid, 3,5-dimethyl-, ethyl ester,3,5-dimethyl-1h-pyrrole-2-carboxylic acidethyl ester,ethyl 3,5-dimethyl-1h-pyrrole-2-carboxylate,pubchem23984,acmc-1cdc7,3,5-dimethyl-1h-pyrrole-2-carboxylicacidethylester,ksc496q7t PubChem CID: 137477 IUPAC Name: ethyl 3,5-dimethyl-1H-pyrrole-2-carboxylate SMILES: CCOC(=O)C1=C(C=C(N1)C)C