CAS RN 21901-29-1
CAS RN 21901-29-1
IUPAC Name:
2-amino-6-methyl-3-nitropyridin-1-ium
Synonyms:
2-amino-6-methyl-3-nitropyridin-1-ium
Molecular Weight (g/mol):
154.15
Molecular Formula:
C6H8N3O2
InChi Key:
LCJXSRQGDONHRK-UHFFFAOYSA-O
SMILES:
CC1=CC=C(C(N)=[NH+]1)[N+]([O-])=O
1
–
2
of
2
results
1
2-Amino-6-methyl-3-nitropyridine, 98%
CAS: 21901-29-1 Molecular Formula: C6H7N3O2 Molecular Weight (g/mol): 153.141 MDL Number: MFCD00047443 InChI Key: LCJXSRQGDONHRK-UHFFFAOYSA-N Synonym: 2-amino-3-nitro-6-picoline,2-amino-6-methyl-3-nitropyridine,2-amino-3-nitro-6-methylpyridine,6-amino-5-nitro-2-picoline,6-methyl-3-nitro-pyridin-2-ylamine,6-methyl-3-nitro-2-pyridinamine,2-pyridinamine, 6-methyl-3-nitro,6-methyl-3-nitro-2-pyridylamine,6-methyl-3-nitropyridin-2-amin,2-amino-3-nitro-6-picoline 2-amino-6-methyl-3-nitropyridine PubChem CID: 226028 IUPAC Name: 6-methyl-3-nitropyridin-2-amine SMILES: CC1=NC(=C(C=C1)[N+](=O)[O-])N