CAS RN 2177270-88-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
12-Aminododecylphosphonic acid hydrochloride, 95%
CAS: 2177270-88-9 Molecular Formula: C12H29ClNO3P Molecular Weight (g/mol): 301.79 MDL Number: MFCD17012689 InChI Key: YCIHZAPKEFUYRI-UHFFFAOYSA-N IUPAC Name: (12-aminododecyl)phosphonic acid hydrochloride SMILES: Cl.NCCCCCCCCCCCCP(O)(O)=O