accessibility menu, dialog, popup

UserName

CAS RN 21510-43-0

BrONN

CAS RN 21510-43-0

IUPAC Name: 2-(4-bromophenyl)-5-phenyl-1,3,4-oxadiazole
Synonyms: 2-(4-bromophenyl)-5-phenyl-1,3,4-oxadiazole
Molecular Weight (g/mol): 301.14
Molecular Formula: C14H9BrN2O
InChi Key: CNLVYZSUMYQALH-UHFFFAOYSA-N
SMILES: BrC1=CC=C(C=C1)C1=NN=C(O1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results