accessibility menu, dialog, popup

UserName

CAS RN 214210-21-6

HOBOHFN

CAS RN 214210-21-6

IUPAC Name: (3-cyano-4-fluorophenyl)boronic acid
Synonyms: 3-cyano-4-fluorophenylboronic acid
Molecular Weight (g/mol): 164.93
Molecular Formula: C7H5BFNO2
InChi Key: OLKIYJDSLMKNLC-UHFFFAOYSA-N
SMILES: OB(O)C1=CC=C(F)C(=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results