CAS RN 21411-42-7
CAS RN 21411-42-7
IUPAC Name:
(2,4,6-trimethylphenyl)methanethiol
Synonyms:
(2,4,6-trimethylphenyl)methanethiol
Molecular Weight (g/mol):
166.28
Molecular Formula:
C10H14S
InChi Key:
LJZYEPXPMRRXTA-UHFFFAOYSA-N
SMILES:
CC1=CC(C)=C(CS)C(C)=C1
1
–
1
of
1
results
1
Alfa Aesar™ 2,4,6-Trimethylbenzyl mercaptan, 97%
CAS: 21411-42-7 Molecular Formula: C10H14S Molecular Weight (g/mol): 166.282 MDL Number: MFCD00173911 InChI Key: LJZYEPXPMRRXTA-UHFFFAOYSA-N Synonym: 2,4,6-trimethylbenzyl mercaptan,mesitylmethanethiol,2,4,6-trimethylphenyl methanethiol,benzenemethanethiol,2,4,6-trimethyl,benzenemethanethiol, 2,4,6-trimethyl,trimethylbenzylmercaptane,acmc-1ch2x,2,4,6-trimethylbenzylmercaptan,2,4,6-trimethylphenyl methane-1-thiol PubChem CID: 301953 IUPAC Name: (2,4,6-trimethylphenyl)methanethiol SMILES: CC1=CC(=C(C(=C1)C)CS)C