CAS RN 2106-18-5
CAS RN 2106-18-5
IUPAC Name:
1-fluoro-2-(trifluoromethoxy)benzene
Synonyms:
1-fluoro-2-(trifluoromethoxy)benzene
Molecular Weight (g/mol):
180.1
Molecular Formula:
C7H4F4O
InChi Key:
UKRYEFFTFFRSPY-UHFFFAOYSA-N
SMILES:
FC1=CC=CC=C1OC(F)(F)F
1
–
1
of
1
results
1
1-Fluoro-2-(trifluoromethoxy)benzene, 97+%, Thermo Scientific™
CAS: 2106-18-5 Molecular Formula: C7H4F4O Molecular Weight (g/mol): 180.102 MDL Number: MFCD00236322 InChI Key: UKRYEFFTFFRSPY-UHFFFAOYSA-N Synonym: 2-trifluoromethoxy fluorobenzene,1-fluoro-2-trifluoromethoxy benzene,2-fluorotrifluoromethoxybenzene,alpha,alpha,alpha,2-tetrafluoroanisole,benzene, fluoro trifluoromethoxy,2-fluorophenyl trifluoromethyl ether,trifluoro 2-fluorophenoxy methane,otf-byl,pubchem10392 PubChem CID: 2777283 IUPAC Name: 1-fluoro-2-(trifluoromethoxy)benzene SMILES: C1=CC=C(C(=C1)OC(F)(F)F)F