CAS RN 209866-92-2
CAS RN 209866-92-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Isoxadifen, TRC
CAS: 209866-92-2 Molecular Formula: C16 H13 N O3 Molecular Weight (g/mol): 267.28 Synonym: Isoxadifen (free acid),3-Isoxazolecarboxylic acid, 4,5-dihydro-5,5-diphenyl- (9CI, ACI),4,5-Dihydro-5,5-diphenyl-3-isoxazolecarboxylic acid (ACI),Isoxadifen IUPAC Name: 5,5-diphenyl-4H-1,2-oxazole-3-carboxylic acid SMILES: OC(=O)C1=NOC(C1)(c2ccccc2)c3ccccc3