accessibility menu, dialog, popup

UserName

CAS RN 2096-10-8

N(R)N(R)O(S)(R)NNOHNH2HOHOH2N

CAS RN 2096-10-8

IUPAC Name: 2-(2,6-diamino-9H-purin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Synonyms: 2-(2,6-diaminopurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol
Molecular Weight (g/mol): 282.26
Molecular Formula: C10H14N6O4
InChi Key: ZDTFMPXQUSBYRL-UHFFFAOYNA-N
SMILES: NC1=NC(N)=C2N=CN(C3OC(CO)C(O)C3O)C2=N1
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results