accessibility menu, dialog, popup

UserName

H3COFFFF

CAS RN 208173-21-1

IUPAC Name: 1-[4-fluoro-2-(trifluoromethyl)phenyl]ethan-1-one
Synonyms: 1-[4-fluoro-2-(trifluoromethyl)phenyl]ethanone
Molecular Weight (g/mol): 206.14
Molecular Formula: C9H6F4O
InChi Key: WBCCAINPZLAKRN-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=C(F)C=C1C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results