accessibility menu, dialog, popup

UserName

CAS RN 207974-14-9

SOOClFBrF

CAS RN 207974-14-9

IUPAC Name: 4-bromo-2,5-difluorobenzene-1-sulfonyl chloride
Synonyms: 4-bromo-2,5-difluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 291.49
Molecular Formula: C6H2BrClF2O2S
InChi Key: SBMKFWMFNIEPDN-UHFFFAOYSA-N
SMILES: FC1=CC(=C(F)C=C1Br)S(Cl)(=O)=O
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Boiling Point 
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)