accessibility menu, dialog, popup

UserName

CAS RN 207801-28-3

CH3OOCH3OOCH3OOY

CAS RN 207801-28-3

IUPAC Name: yttrium(3+) triacetate tetrahydrate
Synonyms: yttrium(3+) triacetate tetrahydrate
Molecular Weight (g/mol): 338.1
Molecular Formula: C6H17O10Y
InChi Key: AIQRTHPXPDTMBQ-UHFFFAOYSA-K
SMILES: O.O.O.O.[Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)