CAS RN 206986-92-7
CAS RN 206986-92-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Cobalt(II) hexafluoro-2,4-pentanedionate hydrate, 97%, Thermo Scientific™
CAS: 206986-92-7 Molecular Formula: C10H6CoF12O5 Molecular Weight (g/mol): 493.07 MDL Number: MFCD00150649,MFCD00042510 InChI Key: POHWVOSIFCILEF-UHFFFAOYSA-N Synonym: bis hexafluoroacetylacetonato cobalt ii hydrate,cobalt 2+ bis 1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate,cobalt 2+ ; z-1,1,1,5,5,5-hexafluoro-4-oxopent-2-en-2-olate PubChem CID: 16212172 IUPAC Name: cobalt;(Z)-1,1,1,5,5,5-hexafluoro-4-hydroxypent-3-en-2-one;hydrate SMILES: O.[Co].OC(=CC(=O)C(F)(F)F)C(F)(F)F.OC(=CC(=O)C(F)(F)F)C(F)(F)F