accessibility menu, dialog, popup

UserName

CAS RN 206559-46-8

BrFFFNCS

CAS RN 206559-46-8

IUPAC Name: 4-bromo-1-isothiocyanato-2-(trifluoromethyl)benzene
Synonyms: 4-bromo-1-isothiocyanato-2-(trifluoromethyl)benzene
Molecular Weight (g/mol): 282.08
Molecular Formula: C8H3BrF3NS
InChi Key: GWLIKBAQYRDIID-UHFFFAOYSA-N
SMILES: FC(F)(F)C1=C(C=CC(Br)=C1)N=C=S
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results