accessibility menu, dialog, popup

UserName

CAS RN 204512-95-8

IUPAC Name: oxolan-3-amine hydrochloride
Synonyms: oxolan-3-amine hydrochloride
Molecular Weight (g/mol): 123.58
Molecular Formula: C4H10ClNO
InChi Key: MHOVLDXJDIEEMJ-UHFFFAOYNA-N
SMILES: Cl.NC1CCOC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results