accessibility menu, dialog, popup

UserName

CAS RN 204387-55-3

CH3HNCH3SOOOH

CAS RN 204387-55-3

IUPAC Name: 2-methylpyrrolidine; 4-methylbenzene-1-sulfonic acid
Synonyms: 2-methylpyrrolidine; para-toluene sulfonate
Molecular Weight (g/mol): 257.35
Molecular Formula: C12H19NO3S
InChi Key: ZXYDRZKLQBDQTN-UHFFFAOYNA-N
SMILES: CC1CCCN1.CC1=CC=C(C=C1)S(O)(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results