accessibility menu, dialog, popup

UserName

CAS RN 20409-48-7

H3CH3CCH3OS

CAS RN 20409-48-7

IUPAC Name: 2,2-dimethyl-1-(thiophen-2-yl)propan-1-one
Synonyms: 2,2-dimethyl-1-(thiophen-2-yl)propan-1-one
Molecular Weight (g/mol): 168.25
Molecular Formula: C9H12OS
InChi Key: PMUOKYKJIKZPIR-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)C1=CC=CS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results