CAS RN 203245-16-3
IUPAC Name:
1,2,3-trifluoro-4-methoxybenzene
Synonyms:
1,2,3-trifluoro-4-methoxybenzene
Molecular Weight (g/mol):
162.11
Molecular Formula:
C7H5F3O
InChi Key:
LIVNSQZJDRPZKP-UHFFFAOYSA-N
SMILES:
COC1=CC=C(F)C(F)=C1F
1
–
1
of
1
results
1
2,3,4-Trifluoroanisole, 97+%, Thermo Scientific™
CAS: 203245-16-3 Molecular Formula: C7H5F3O Molecular Weight (g/mol): 162.111 MDL Number: MFCD00042396 InChI Key: LIVNSQZJDRPZKP-UHFFFAOYSA-N Synonym: 2,3,4-trifluoroanisole,benzene, 1,2,3-trifluoro-4-methoxy-9ci,benzene, 1,2,3-trifluoro-4-methoxy,acmc-20alqi,methyl 2,3,4-trifluorophenyl ether,1,2,3-trifluoro-4-methoxy-benzene,2,3,4-trifluorophenol, methyl ether,1-methoxy-2,3,4-trifluorobenzene PubChem CID: 2776942 IUPAC Name: 1,2,3-trifluoro-4-methoxybenzene SMILES: COC1=C(C(=C(C=C1)F)F)F