accessibility menu, dialog, popup

UserName

CAS RN 20289-26-3

OHN

CAS RN 20289-26-3

IUPAC Name: 4-(benzyloxy)-1H-indole
Synonyms: 4-(benzyloxy)-1H-indole
Molecular Weight (g/mol): 223.28
Molecular Formula: C15H13NO
InChi Key: LJFVSIDBFJPKLD-UHFFFAOYSA-N
SMILES: C(OC1=C2C=CNC2=CC=C1)C1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results