CAS RN 2028-12-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-(Aminocarbonyl)-3-cyclohexene-1-carboxylic Acid, TRC
CAS: 2028-12-8 Molecular Formula: C8 H11 N O3 Molecular Weight (g/mol): 169.178 Synonym: 3-Cyclohexene-1-carboxylic acid, 6-carbamoyl- (6CI,7CI,8CI),6-(Aminocarbonyl)-3-cyclohexene-1-carboxylic acid,6-Carbamoylcyclohex-3-enecarboxylic acid,NSC 191910 IUPAC Name: 6-carbamoylcyclohex-3-ene-1-carboxylic acid SMILES: NC(=O)C1CC=CCC1C(=O)O