CAS RN 20208-95-1
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Melamine Phosphate, TRC
CAS: 20208-95-1 Molecular Formula: C3 H6 N6 . H3 O4 P Molecular Weight (g/mol): 224.115 Synonym: 1,3,5-Triazine-2,4,6-triamine, phosphate (1:1),Melamine, phosphate (1:1) (8CI),1,3,5-Triazinane-2,4,6-triimine phosphate,Amgard NH,M 850103,Melagard MP,Melamine monophosphate,Melamine phosphate IUPAC Name: phosphoric acid;1,3,5-triazine-2,4,6-triamine SMILES: Nc1nc(N)nc(N)n1.OP(=O)(O)O