accessibility menu, dialog, popup

UserName

CAS RN 202001-00-1

OOFCl

CAS RN 202001-00-1

IUPAC Name: 2-(3-chloro-5-fluorophenyl)acetate
Synonyms: 2-(3-chloro-5-fluorophenyl)acetate
Molecular Weight (g/mol): 187.57
Molecular Formula: C8H5ClFO2
InChi Key: GFLQMGYDUPLXHZ-UHFFFAOYSA-M
SMILES: [O-]C(=O)CC1=CC(F)=CC(Cl)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results