accessibility menu, dialog, popup

UserName

CAS RN 20109-09-5

HO

CAS RN 20109-09-5

IUPAC Name: 1-(2-phenylethynyl)cyclohexan-1-ol
Synonyms: 1-(2-phenylethynyl)cyclohexan-1-ol
Molecular Weight (g/mol): 200.28
Molecular Formula: C14H16O
InChi Key: LBEMYSKBSYEVAG-UHFFFAOYSA-N
SMILES: OC1(CCCCC1)C#CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results