CAS RN 20029-52-1
IUPAC Name:
4-cyclohexylbenzoic acid
Synonyms:
4-cyclohexylbenzoic acid
Molecular Weight (g/mol):
204.27
Molecular Formula:
C13H16O2
InChi Key:
QCIWHVKGVVQHIY-UHFFFAOYSA-N
SMILES:
OC(=O)C1=CC=C(C=C1)C1CCCCC1
1
–
1
of
1
results
1
4-Cyclohexylbenzoic acid, 98%
CAS: 20029-52-1 Molecular Formula: C13H16O2 Molecular Weight (g/mol): 204.269 MDL Number: MFCD00039459 InChI Key: QCIWHVKGVVQHIY-UHFFFAOYSA-N Synonym: 4-cyclohexyl-benzoic acid,benzoic acid, 4-cyclohexyl,4cyclohexylbenzoic acid,4-cyclohexylbenzoicacid,p-cyclohexylbenzoic acid,acmc-1cmbu,cyclohexybenzoicacid,4-carboxyphenyl cyclohexane PubChem CID: 88337 IUPAC Name: 4-cyclohexylbenzoic acid SMILES: C1CCC(CC1)C2=CC=C(C=C2)C(=O)O