accessibility menu, dialog, popup

UserName

ClClO

CAS RN 19811-05-3

IUPAC Name: (2,4-dichlorophenyl)(phenyl)methanone
Synonyms: (2,4-dichlorophenyl)(phenyl)methanone
Molecular Weight (g/mol): 251.11
Molecular Formula: C13H8Cl2O
InChi Key: VLTYTTRXESKBKI-UHFFFAOYSA-N
SMILES: ClC1=CC=C(C(=O)C2=CC=CC=C2)C(Cl)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results