accessibility menu, dialog, popup

UserName

CAS RN 19780-59-7

H3CCH3CH3H3COH

CAS RN 19780-59-7

IUPAC Name: 3-ethyl-2-methylheptan-2-ol
Synonyms: 3-ethyl-2-methylheptan-2-ol
Molecular Weight (g/mol): 158.29
Molecular Formula: C10H22O
InChi Key: IDMQBLFACWMOGD-UHFFFAOYNA-N
SMILES: CCCCC(CC)C(C)(C)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results