accessibility menu, dialog, popup

UserName

H3CCH3H3COOOHNCH3OO

CAS RN 197632-77-2

IUPAC Name: 3-(tert-butoxy)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}(methyl)amino)propanoic acid
Synonyms: 3-(tert-butoxy)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl](methyl)amino}propanoic acid
Molecular Weight (g/mol): 397.47
Molecular Formula: C23H27NO5
InChi Key: PQSAXALAXPNFMG-UHFFFAOYNA-N
SMILES: CN(C(COC(C)(C)C)C(O)=O)C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results