accessibility menu, dialog, popup

UserName

CAS RN 195622-41-4

CH3OFFFOOH

CAS RN 195622-41-4

IUPAC Name: 2-methyl-5-(trifluoromethoxy)benzoic acid
Synonyms: 2-methyl-5-(trifluoromethoxy)benzoic acid
Molecular Weight (g/mol): 220.15
Molecular Formula: C9H7F3O3
InChi Key: IBHIJPBIQCIAES-UHFFFAOYSA-N
SMILES: CC1=C(C=C(OC(F)(F)F)C=C1)C(O)=O
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results