CAS RN 19506-87-7
CAS RN 19506-87-7
IUPAC Name:
2-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid
Synonyms:
2-(1,3-dioxoisoindol-2-yl)propanoic acid
Molecular Weight (g/mol):
219.2
Molecular Formula:
C11H9NO4
InChi Key:
OZWUITKBAWTEAQ-UHFFFAOYNA-N
SMILES:
CC(N1C(=O)C2=CC=CC=C2C1=O)C(O)=O
1
–
1
of
1
results
1
2-Phthalimidopropionic acid, 98%, Thermo Scientific™
CAS: 19506-87-7 Molecular Formula: C11H9NO4 Molecular Weight (g/mol): 219.196 MDL Number: MFCD00023061 InChI Key: OZWUITKBAWTEAQ-UHFFFAOYSA-N Synonym: 2-phthalimidopropionic acid,phthaloyl-dl-alanine,2-1,3-dioxo-1,3-dihydro-2h-isoindol-2-yl propanoic acid,2-1,3-dioxoisoindolin-2-yl propanoic acid,phthalyl-dl-alanine,2-1,3-dioxoisoindol-2-yl propanoic acid,2-1,3-dihydro-1,3-dioxo-2h-isoindol-2-yl propanoic acid,2-phthalimidopropanoic acid,2-1,3-dioxo-2,3-dihydro-1h-isoindol-2-yl propanoic acid,2h-isoindole-2-acetic acid, 1,3-dihydro-alpha-methyl-1,3-dioxo PubChem CID: 86884 IUPAC Name: 2-(1,3-dioxoisoindol-2-yl)propanoic acid SMILES: CC(C(=O)O)N1C(=O)C2=CC=CC=C2C1=O